Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir52b
- Entrez Gene ID
-
246631
- Gene Name
- Ionotropic receptor 52b
- Gene Aliases
-
CG12961, CG30469, DmelIR52b, Dmel\CG30469, Dmel_CG30469, IF52b, IR52b
- Chromosome Location
-
Chr.2R: 15501820 - 15503610 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050469
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166097.2 |
NP_725469.2 |
1 - 1 |
1658 |
80 |
View Full Product Details