Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Flo1
Entrez Gene ID
36726
Gene Name
Flotillin 1
Gene Aliases
CG8200, CK02126, Dmel\CG8200, Dmel_CG8200, FLO1, FLO[Dm], FLO[Dm]-1, Flo, Flo-1, Flo[Dm], dFlot1, flo, flo1, flotillin, reggie-2
Chromosome Location
Chr.2R: 15566271 - 15569404 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024754
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058010.4 NP_477358.1 5 - 6 1334 85
NM_166102.2 NP_725476.1 6 - 7 1346 85
GenBank mRNA AF044734.1 - 5 - 6 1100 85

View Full Product Details