Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bdg
Entrez Gene ID
36747
Gene Name
bedraggled
Gene Aliases
CG8291, Dmel\CG8291, Dmel_CG8291
Chromosome Location
Chr.2R: 15923040 - 15931935 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034049
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299540.1 NP_001286469.1 2 - 3 1086 72
NM_137220.3 NP_611064.2 2 - 3 1086 72
NM_166121.2 NP_725499.2 1 - 2 1593 72
NM_176181.2 NP_788361.1 2 - 3 1299 72
GenBank mRNA BT150416.1 - 1 - 2 1593 72

View Full Product Details