Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spin
Entrez Gene ID
45380
Gene Name
spinster
Gene Aliases
25/11, CG8428, Dmel\CG8428, Dmel_CG8428, Spin, bnch, l(2)10403, l(2)W5, l(2)k02511, l(2)k09905, l(2R)W5
Chromosome Location
Chr.2R: 16124981 - 16138238 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086676
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080084.3 NP_524823.1 7 - 8 2164 95
NM_166146.2 NP_725532.1 7 - 8 2164 95
NM_166147.2 NP_725533.1 6 - 7 1902 95
GenBank mRNA AF212366.1 - 7 - 8 2127 95
AF212367.1 - 7 - 8 2127 95

View Full Product Details