Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
park
Entrez Gene ID
40336
Gene Name
parkin
Gene Aliases
AAN12155.1, CG10523, Dmel\CG10523, Dmel_CG10523, Dpark, PARK, PARK2, PARKIN, PK, Park, Parkin, SD01679, dPARKIN, dParkin, dpark, dpk, park1, pk
Chromosome Location
Chr.3L: 21194754 - 21196853 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041100
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_168884.2 NP_730600.1 6 - 7 1428 67
NM_168885.3 NP_730601.1 5 - 6 1407 67
GenBank mRNA AY093423.3 - 5 - 6 1349 67
BT044504.1 - 5 - 6 1349 67

View Full Product Details