Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hr78
Entrez Gene ID
40378
Gene Name
Hormone-receptor-like in 78
Gene Aliases
CG7199, DHR78, Dhr78, Dmel\CG7199, Dmel_CG7199, HR78, Hr78D, NR2D1, XR78E/F
Chromosome Location
Chr.3L: 21494019 - 21497494 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015239
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202222.1 NP_001189151.1 4 - 5 718 111
NM_001202223.1 NP_001189152.1 3 - 4 711 111
NM_079479.5 NP_524203.2 4 - 5 875 111
NM_168907.3 NP_730636.1 4 - 5 851 111
NM_168908.3 NP_730637.1 4 - 5 896 111
NM_206415.3 NP_996137.1 4 - 5 881 111
GenBank mRNA AY058317.1 - 4 - 5 844 111

View Full Product Details