Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- dlp
- Entrez Gene ID
-
39596
- Gene Name
- dally-like
- Gene Aliases
-
CG17703, CG32146, CG5031, CT16138, D-gpcB, DLP, Dally-like, Dlp, Dly, Dmel\CG32146, Dmel_CG32146, dly
- Chromosome Location
-
Chr.3L: 14632601 - 14671609 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041604
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079347.3 |
NP_524071.2 |
7 - 8 |
2326 |
71 |
GenBank mRNA |
AY052017.1 |
- |
7 - 8 |
2307 |
71 |
View Full Product Details