Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trl
Entrez Gene ID
2768981
Gene Name
Trithorax-like
Gene Aliases
Adf-2, Adf-2-519, Adf2, CG33261, CG9343, Dmel\CG33261, Dmel_CG33261, E(var)3-trl, E(var)62, GAF, GAGA, GAGA factor, Gaf, Gaga, LBC, NC70F, Nc70F, TRL, TfGAGA/Adf-2, Trl-GAGA, anon-EST:fe2E12, gaga, l(3)s2325, trl
Chromosome Location
Chr.3L: 14747929 - 14761049 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013263
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038924.4 NP_001034013.2 2 - 3 897 87
NM_001038925.3 NP_001034014.1 3 - 4 946 87
NM_001038926.2 NP_001034015.1 3 - 4 1012 87
NM_001274917.1 NP_001261846.1 3 - 4 1012 87
NM_206355.4 NP_996077.1 3 - 4 1012 87
NM_206356.3 NP_996078.1 3 - 4 1012 87
NM_206357.3 NP_996079.1 3 - 4 1370 87
NM_206358.4 NP_996080.1 3 - 4 1187 87
NM_206359.4 NP_996081.1 3 - 4 1191 87
NM_206360.1 NP_996082.1 4 - 5 1315 87
GenBank mRNA AY069651.1 - 3 - 4 1170 87

Gene Symbol
CG42507
Entrez Gene ID
8674092
Gene Name
CG42507-PA
Gene Aliases
Dmel\CG42507, Dmel_CG42507
Chromosome Location
Chr.3L: 14747929 - 14757962 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260233
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169972.2 NP_001163443.1 3 - 4 1012 87
NM_001169973.2 NP_001163444.1 3 - 4 1012 87

View Full Product Details