Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CrebA
Entrez Gene ID
39682
Gene Name
Cyclic-AMP response element binding protein A
Gene Aliases
BBF-2, BBF2_DROME, BOX B Binding Factor-2, Bbbf2, BcDNA:SD05937, CG7450, CRE-BP, CREB, CREB-A, CREB-a, CREBA, Creb, Creb A, Creb-A, DM4, Dmel\CG7450, Dmel_CG7450, creb, crebA, dCREB-A, dCREBA, dCreb-A, dCrebA, l(3)03576
Chromosome Location
Chr.3L: 15535395 - 15553214 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004396
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079363.5 NP_524087.3 1 - 2 1057 86
NM_206374.2 NP_996096.1 1 - 2 1057 86
GenBank mRNA BT025219.1 - 1 - 2 1055 86

View Full Product Details