The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144669.2 | NP_001138141.1 | 3 - 4 | 1733 | 94 |
NM_001297813.1 | NP_001284742.1 | 4 - 5 | 1608 | 94 | |
NM_001297814.1 | NP_001284743.1 | 3 - 4 | 1367 | 94 | |
NM_001297815.1 | NP_001284744.1 | 3 - 4 | 1367 | 94 | |
NM_080293.4 | NP_525032.2 | 4 - 5 | 1646 | 94 | |
NM_166846.5 | NP_726675.3 | 3 - 4 | 1660 | 94 | |
NM_176679.3 | NP_788852.2 | 3 - 4 | 1367 | 94 | |
NM_206596.2 | NP_996319.1 | 4 - 5 | 1646 | 94 | |
NM_206597.2 | NP_996320.1 | 4 - 5 | 1646 | 94 | |
NM_206598.3 | NP_996321.1 | 3 - 4 | 1660 | 94 | |
NM_206599.2 | NP_996322.1 | 3 - 4 | 1660 | 94 | |
GenBank mRNA | AF545816.1 | - | 3 - 4 | 1304 | 94 |
AF545817.1 | - | 3 - 4 | 1424 | 94 | |
AF545818.1 | - | 3 - 4 | 1424 | 94 | |
AF545819.1 | - | 3 - 4 | 1304 | 94 | |
AF545820.1 | - | 3 - 4 | 1424 | 94 | |
BT015253.1 | - | 3 - 4 | 1303 | 94 | |
BT099720.1 | - | 3 - 4 | 1337 | 94 | |
BT100053.1 | - | 3 - 4 | 1338 | 94 | |
BT100310.1 | - | 3 - 4 | 846 | 94 |