Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnp4F
Entrez Gene ID
31448
Gene Name
RNA-binding protein 4F
Gene Aliases
4F-rnp, 4f-rnp, CG3312, Dmel\CG3312, Dmel_CG3312, RNP-4F, Rnp-4F, rnp-4f
Chromosome Location
Chr.X: 5318928 - 5322696 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014024
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103413.3 NP_001096883.3 7 - 8 2458 65
NM_001103414.3 NP_001096884.1 8 - 9 2488 65
NM_078492.4 NP_511047.2 7 - 8 2665 65
NM_206630.3 NP_996353.1 8 - 9 2584 65
GenBank mRNA AY061395.1 - 7 - 8 2679 65

View Full Product Details