Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Marf
Entrez Gene ID
31581
Gene Name
Mitochondrial assembly regulatory factor
Gene Aliases
CG3869, CG38869, Dmel\CG3869, Dmel_CG3869, MARF, MFN, MFN2, Marf-1, Mfn, Mfn-2, Mfn2, anon-WO0125274.3, dMFN, dMfn, dMfn2, dmfn, marf, mfn, mfn2
Chromosome Location
Chr.X: 6365949 - 6370924 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029870
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132092.3 NP_572320.2 3 - 4 1103 57
NM_206634.2 NP_996357.1 3 - 4 1103 57
NM_206635.3 NP_996358.2 3 - 4 942 57
GenBank mRNA BT010027.1 - 3 - 4 1103 57

View Full Product Details