Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHES-1-like
Entrez Gene ID
31678
Gene Name
Checkpoint suppressor 1-like
Gene Aliases
CG12690, Ches-1-like, Ches1, Dmches-1, Dmel\CG12690, Dmel_CG12690, FoxN, anon-EST:Posey7, ches1
Chromosome Location
Chr.X: 7682338 - 7709385 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029504
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272382.1 NP_001259311.1 2 - 3 1738 79
NM_001272383.1 NP_001259312.1 2 - 3 1996 79
NM_078516.3 NP_511071.3 2 - 3 1833 79
NM_206639.3 NP_996362.2 2 - 3 1791 79

View Full Product Details