Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
otu
Entrez Gene ID
31789
Gene Name
ovarian tumor
Gene Aliases
CG12743, DROOTUA, Dmel\CG12743, Dmel_CG12743, K, OTU, Otu, fs(1)231, fs(1)23l, fs(1)M101, fs(1)jA265, fs(1)otu
Chromosome Location
Chr.X: 8485008 - 8489535 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003023
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272403.1 NP_001259332.1 5 - 6 952 62
NM_001272404.1 NP_001259333.1 5 - 6 1064 62
NM_078534.4 NP_511089.2 5 - 6 1047 62
NM_167158.3 NP_727271.1 5 - 6 1047 62
NM_206656.2 NP_996379.1 5 - 6 1061 62
GenBank mRNA BT133210.1 - 5 - 6 1012 62

View Full Product Details