Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhp
Entrez Gene ID
32531
Gene Name
Rhophilin
Gene Aliases
CG8497, Dmel\CG8497, Dmel_CG8497
Chromosome Location
Chr.X: 15788897 - 15793632 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026374
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078613.4 NP_511168.3 4 - 5 451 73
NM_206724.2 NP_996447.1 4 - 5 606 73
GenBank mRNA AF132025.1 - 4 - 5 455 73
BT021401.1 - 4 - 5 455 73

View Full Product Details