Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
shi
Entrez Gene ID
45928
Gene Name
shibire
Gene Aliases
CG18102, DNM3, DYN, Ddyn, Ddyn3, Ddyn4, Dmel\CG18102, Dmel_CG18102, Dyn, Shi, Shibire, Vps1, anon-WO0153538.12, anon-WO0153538.13, anon-WO0153538.14, dDyn, dyn, dynamin, l(1)VII, l(1)shi, shibere, shu
Chromosome Location
Chr.X: 15892116 - 15906716 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003392
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169295.1 NP_001162766.1 14 - 15 2906 87
NM_001169296.1 NP_001162767.1 13 - 14 2622 87
NM_001272659.2 NP_001259588.1 15 - 16 2774 87
NM_001298377.1 NP_001285306.1 15 - 16 2729 87
NM_167470.3 NP_727910.1 14 - 15 2826 87
NM_206742.1 NP_996465.1 14 - 15 2716 87
NM_206743.2 NP_996466.1 14 - 15 2717 87
GenBank mRNA BT010049.1 - 14 - 15 2883 87

View Full Product Details