Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Lcch3
- Entrez Gene ID
-
32554
- Gene Name
- Ligand-gated chloride channel homolog 3
- Gene Aliases
-
CG17336, CT38330, DmGABAbeta, DmGA_b, Dmel\CG17336, Dmel_CG17336, LCCH3
- Chromosome Location
-
Chr.X: 15925105 - 15928529 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010240
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206746.2 |
NP_996469.1 |
7 - 8 |
1604 |
74 |
GenBank mRNA |
AY060660.1 |
- |
6 - 7 |
2409 |
74 |
View Full Product Details