Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mmd
Entrez Gene ID
32561
Gene Name
mind-meld
Gene Aliases
CG15603, CG15604, CG42252, CG9163, CT26192, Dmel\CG42252, Dmel_CG42252
Chromosome Location
Chr.X: 15946936 - 15973159 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259110
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144728.3 NP_001138200.1 6 - 7 1275 74
NM_001258775.3 NP_001245704.1 6 - 7 1275 74
NM_001258776.2 NP_001245705.1 7 - 8 1266 74
NM_001272661.2 NP_001259590.1 7 - 8 1474 74
NM_001272662.2 NP_001259591.1 7 - 8 1474 74
NM_001298382.1 NP_001285311.1 6 - 7 1957 74
NM_078634.7 NP_523358.5 6 - 7 1957 74
NM_206752.3 NP_996475.2 6 - 7 1275 74

View Full Product Details