Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mmd
Entrez Gene ID
32561
Gene Name
mind-meld
Gene Aliases
CG15603, CG15604, CG42252, CG9163, CT26192, Dmel\CG42252, Dmel_CG42252
Chromosome Location
Chr.X: 15946936 - 15973159 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259110
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144728.3 NP_001138200.1 10 - 11 1967 91
NM_001258775.3 NP_001245704.1 10 - 11 1967 91
NM_001258776.2 NP_001245705.1 11 - 12 1958 91
NM_001272661.2 NP_001259590.1 11 - 12 2166 91
NM_001272662.2 NP_001259591.1 11 - 12 2166 91
NM_001298382.1 NP_001285311.1 10 - 11 2649 91
NM_078634.7 NP_523358.5 10 - 11 2649 91
NM_206752.3 NP_996475.2 10 - 11 1967 91

View Full Product Details