Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eIF5
- Entrez Gene ID
-
32566
- Gene Name
- eukaryotic translation initiation factor 5
- Gene Aliases
-
CG9177, Dmel\CG9177, Dmel_CG9177, EIf5, anon-EST:fe3C6
- Chromosome Location
-
Chr.X: 15987008 - 15991974 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030719
- Gene Symbol
- CG46312
- Entrez Gene ID
-
30522401
- Gene Name
- CG46312-PA
- Gene Aliases
-
Dmel\CG46312, Dmel_CG46312, eIF5-5'utr
- Chromosome Location
-
Chr.X: 15987166 - 15991974 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0284228
View Full Product Details