Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh
Entrez Gene ID
32780
Gene Name
Shaker
Gene Aliases
BcDNA:GH03046, CG12348, CG17860, CG7640, Dmel\CG12348, Dmel_CG12348, EKO, Eko, F-c, K[[v]]1, Kv1, SH, Sha, Shw, V-I, minisleep, sh, sha
Chromosome Location
Chr.X: 17924307 - 18063247 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003380
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272736.2 NP_001259665.2 2 - 3 720 94
NM_001316640.1 NP_001303569.1 2 - 3 785 94
NM_078669.5 NP_523393.3 2 - 3 785 94
NM_167595.4 NP_728123.1 2 - 3 785 94
NM_206774.2 NP_996497.1 2 - 3 728 94
GenBank mRNA X07134.1 - - 728 94

View Full Product Details