Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sh
Entrez Gene ID
32780
Gene Name
Shaker
Gene Aliases
BcDNA:GH03046, CG12348, CG17860, CG7640, Dmel\CG12348, Dmel_CG12348, EKO, Eko, F-c, K[[v]]1, Kv1, SH, Sha, Shw, V-I, minisleep, sh, sha
Chromosome Location
Chr.X: 17924307 - 18063247 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003380
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169317.1 NP_001162788.1 6 - 7 949 124
NM_001201739.2 NP_001188668.1 8 - 9 3241 124
NM_001258797.1 NP_001245726.1 7 - 8 1884 124
NM_001258798.2 NP_001245727.1 10 - 11 2848 124
NM_001272735.3 NP_001259664.1 8 - 9 3241 124
NM_001272736.2 NP_001259665.2 7 - 8 1429 124
NM_001272737.1 NP_001259666.2 7 - 8 2204 124
NM_001316640.1 NP_001303569.1 7 - 8 1494 124
NM_078669.5 NP_523393.3 7 - 8 1494 124
NM_167592.4 NP_728120.1 7 - 8 1028 124
NM_167594.4 NP_728122.1 8 - 9 3241 124
NM_167595.4 NP_728123.1 7 - 8 1494 124
NM_167596.5 NP_728124.1 7 - 8 1028 124
NM_206774.2 NP_996497.1 7 - 8 1437 124
NM_206775.5 NP_996498.2 10 - 11 2848 124
GenBank mRNA BT050432.1 - 7 - 8 1028 124
X06184.1 - - 947 124
X07132.1 - - 1122 124
X07133.1 - - 826 124
X07134.1 - - 1437 124

View Full Product Details