Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpt6
Entrez Gene ID
33105
Gene Name
Regulatory particle triple-A ATPase 6
Gene Aliases
1489, CG1489, DUG, Dm_Rpt6a, Dmel\CG1489, Dmel_CG1489, Dug, PRS8_DROME, Pros45, S8, Ug, dRpt6, p42C, pros45, sug-1, sug1
Chromosome Location
Chr.X: 21316052 - 21317663 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020369
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134603.2 NP_608447.1 2 - 3 295 81
GenBank mRNA AF043734.1 - 2 - 3 232 81
AY051732.1 - 2 - 3 233 81

View Full Product Details