Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rempA
- Entrez Gene ID
-
33230
- Gene Name
- reduced mechanoreceptor potential A
- Gene Aliases
-
CG11838, CHE-11, Che-11, Dmel\CG11838, Dmel_CG11838, IFT140, IFT40, OSEG, OSEG3, Oseg3, REMPA, RempA, oseg3, remp
- Chromosome Location
-
Chr.2L: 553742 - 560310 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0260933
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205886.2 |
NP_995608.1 |
1 - 2 |
463 |
66 |
View Full Product Details