Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tspo
- Entrez Gene ID
-
33231
- Gene Name
- Translocator protein
- Gene Aliases
-
CG2789, Dmel\CG2789, Dmel_CG2789, anon-EST:Posey116, dTSPO, tspo
- Chromosome Location
-
Chr.2L: 559612 - 560523 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031263
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134687.4 |
NP_608531.1 |
1 - 2 |
426 |
55 |
GenBank mRNA |
AY058269.1 |
- |
1 - 2 |
341 |
55 |
View Full Product Details