Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frtz
Entrez Gene ID
33349
Gene Name
fritz
Gene Aliases
CG17657, Dmel\CG17657, Dmel_CG17657, anon-WO0118547.105
Chromosome Location
Chr.2L: 1711302 - 1715851 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086698
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272950.1 NP_001259879.1 5 - 6 2870 87
NM_001272951.1 NP_001259880.1 5 - 6 2855 87
NM_134770.2 NP_608614.1 5 - 6 2870 87
GenBank mRNA BT004832.1 - 5 - 6 2882 87

View Full Product Details