Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mio
- Entrez Gene ID
-
33399
- Gene Name
- missing oocyte
- Gene Aliases
-
CG15384, CG7074, Dmel\CG7074, Dmel_CG7074, FBtr0077835, Mio, SEA4, Sea4
- Chromosome Location
-
Chr.2L: 2217290 - 2220625 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031399
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134812.3 |
NP_608656.1 |
4 - 5 |
1604 |
80 |
GenBank mRNA |
AY061485.1 |
- |
4 - 5 |
1531 |
80 |
View Full Product Details