The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272993.1 | NP_001259922.1 | 2 - 2 | 120 | 81 |
NM_134823.3 | NP_608667.1 | 1 - 1 | 141 | 81 | |
GenBank mRNA | BT023397.1 | - | 1 - 1 | 116 | 81 |
BT029393.1 | - | 1 - 1 | 116 | 81 | |
BT029394.1 | - | 1 - 1 | 116 | 81 | |
KX671610.1 | - | 1 - 1 | 65 | 81 |