Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnpo-SR
Entrez Gene ID
33465
Gene Name
Transportin-Serine/Arginine rich
Gene Aliases
CG2848, Dmel\CG2848, Dmel_CG2848, TNPO3, Trn-SR, anon-EST:Posey294, dTRN-SR
Chromosome Location
Chr.2L: 2759924 - 2763889 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031456
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273032.1 NP_001259961.1 3 - 4 1479 85
NM_134864.4 NP_608708.2 3 - 4 1479 85
GenBank mRNA AY095186.1 - 3 - 4 1434 85

View Full Product Details