Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Spindly
- Entrez Gene ID
-
33578
- Gene Name
- CG15415-PB
- Gene Aliases
-
CG15415, Dmel\CG15415, Dmel_CG15415, spindly
- Chromosome Location
-
Chr.2L: 3527387 - 3530317 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031549
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134947.2 |
NP_608791.2 |
3 - 4 |
925 |
66 |
View Full Product Details