Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mfr
Entrez Gene ID
39016
Gene Name
misfire
Gene Aliases
CG5747, Dmel\CG5747, Dmel_CG5747, Otoferlin
Chromosome Location
Chr.3L: 8905918 - 8912671 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266757
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144447.2 NP_001137919.1 3 - 4 796 78

View Full Product Details