Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
teq
Entrez Gene ID
39048
Gene Name
Tequila
Gene Aliases
BEST:CK00198, BEST:GH18038, CG18153, CG18403, CG4821, CG4948, CK00198, Dmel\CG4821, Dmel_CG4821, SP72, Teq, anon-EST:ParkEST161, graal
Chromosome Location
Chr.3L: 9074643 - 9092131 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023479
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169922.3 NP_001163393.2 8 - 9 2458 91
NM_140031.3 NP_648288.2 8 - 9 2458 91

View Full Product Details