Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GlcAT-P
Entrez Gene ID
39262
Gene Name
Glucuronyltransferase P
Gene Aliases
CG6207, DGlcAT-P, DmGlcAT-BSII, Dmel\CG6207, Dmel_CG6207, dGlcAT-BSII, dGlcAT-P
Chromosome Location
Chr.3L: 11176954 - 11197536 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036144
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014581.3 NP_001014581.1 5 - 6 1240 86
NM_001259784.2 NP_001246713.1 6 - 7 1628 86
NM_001259785.2 NP_001246714.1 5 - 6 1135 86
NM_140191.3 NP_648448.1 6 - 7 1729 86
NM_168451.4 NP_729685.2 5 - 6 1401 86
GenBank mRNA AB080697.1 - 2 - 3 470 86
AY118925.1 - 5 - 6 1207 86
BT021381.1 - 5 - 6 1355 86

View Full Product Details