Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ir68a
Entrez Gene ID
39269
Gene Name
Ionotropic receptor 68a
Gene Aliases
CG6185, CT19410, DmelIR68a, Dmel\CG6185, Dmel_CG6185, IR68a
Chromosome Location
Chr.3L: 11232972 - 11236218 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036150
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300102.1 NP_001287031.1 2 - 3 939 74
NM_140198.3 NP_648455.2 2 - 3 939 74

View Full Product Details