Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- obst-G
- Entrez Gene ID
-
39355
- Gene Name
- obstructor-G
- Gene Aliases
-
CG9781, Dmel\CG9781, Dmel_CG9781
- Chromosome Location
-
Chr.3L: 11952301 - 11953317 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036228
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140272.2 |
NP_648529.1 |
1 - 2 |
84 |
82 |
View Full Product Details