Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tango1
Entrez Gene ID
33930
Gene Name
Transport and Golgi organization 1
Gene Aliases
2L3443, CG11098, Dmel\CG11098, Dmel_CG11098, TANGO1, tango1
Chromosome Location
Chr.2L: 6649388 - 6654574 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0286898
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135214.4 NP_609058.2 3 - 4 2845 84
NM_164697.3 NP_723198.2 3 - 4 2845 84
GenBank mRNA BT003314.1 - 3 - 4 2805 84

View Full Product Details