Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
milt
Entrez Gene ID
45683
Gene Name
milton
Gene Aliases
CG13777, CG31630, CG43227, Dmel\CG43227, Dmel_CG43227, Milt, Milton, anon-WO0153538.19, l(2)k04704
Chromosome Location
Chr.2L: 7037823 - 7056010 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262872
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080765.4 NP_542943.2 2 - 3 849 74
NM_164736.3 NP_723249.1 2 - 3 761 74
GenBank mRNA BT133374.1 - 2 - 3 516 74

View Full Product Details