Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RapGAP1
Entrez Gene ID
34032
Gene Name
Rap GTPase activating protein 1
Gene Aliases
CG13789, CG13790, CG13791, CG33529, CG34374, CG44086, CG6682, Dmel\CG44086, Dmel_CG44086, FBpp0111515, Q9VM02_DROME, RapGAP, RapGap1, Rapgap1, rapgap, rapgap1
Chromosome Location
Chr.2L: 7497805 - 7578980 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264895
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014478.2 NP_001014478.1 2 - 3 3081 76
NM_001014479.2 NP_001014479.1 3 - 4 2352 76
NM_001103644.3 NP_001097114.2 3 - 4 2651 76
NM_001273279.1 NP_001260208.1 4 - 5 2592 76
NM_144299.3 NP_652556.2 4 - 5 2675 76

View Full Product Details