Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tep2
- Entrez Gene ID
-
34044
- Gene Name
- Thioester-containing protein 2
- Gene Aliases
-
145970_at, 145971_at, CG18589, CG7052, Dmel\CG7052, Dmel_CG7052, T13, TEP II, TEP2, TEP2mel, Tep 2, Tep II, TepII, dTEPII, tepII
- Chromosome Location
-
Chr.2L: 7693727 - 7701601 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041182
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_164761.2 |
NP_723298.1 |
5 - 6 |
2214 |
129 |
View Full Product Details