Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Herp
Entrez Gene ID
34065
Gene Name
Homocysteine-induced endoplasmic reticulum protein
Gene Aliases
CG14536, Dmel\CG14536, Dmel_CG14536, dHerp, herp
Chromosome Location
Chr.2L: 7796314 - 7800058 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031950
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135307.3 NP_609151.2 2 - 3 1204 71
NM_164772.2 NP_723316.1 2 - 3 993 71
GenBank mRNA BT012479.1 - 2 - 3 963 71

View Full Product Details