Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mur29B
- Entrez Gene ID
-
319014
- Gene Name
- Mucin related 29B
- Gene Aliases
-
BcDNA:LP08232, CG31901, Dmel\CG31901, Dmel_CG31901
- Chromosome Location
-
Chr.2L: 8352187 - 8355172 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051901
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_164808.4 |
NP_723377.1 |
2 - 3 |
357 |
91 |
View Full Product Details