Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eaat1
Entrez Gene ID
34251
Gene Name
Excitatory amino acid transporter 1
Gene Aliases
AF001784, CG3747, Dglt-1, Dglut-1, Dmel\CG3747, Dmel_CG3747, EAAT, EAAT1, Eaat 1, dEAAT, dEAAT1, dEaat1, eaat1
Chromosome Location
Chr.2L: 9333567 - 9341925 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026439
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201822.2 NP_001188751.1 3 - 4 663 65
NM_001382081.1 NP_001369122.1 3 - 4 691 65
NM_001382082.1 NP_001369123.1 3 - 4 691 65
NM_058080.4 NP_477428.1 3 - 4 691 65
NM_164853.3 NP_723454.1 3 - 4 495 65
NM_164854.3 NP_723455.1 3 - 4 557 65
GenBank mRNA AF165999.1 - 2 - 3 399 65
BT044100.1 - 3 - 4 548 65
D86739.2 - 3 - 4 668 65

View Full Product Details