Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GlcAT-S
Entrez Gene ID
34282
Gene Name
Glucuronyltransferase S
Gene Aliases
CG3881, DGlcAT-S, DmGlcAT-BSI, Dmel\CG3881, Dmel_CG3881, dGlcAT-BSI, dGlcAT-S
Chromosome Location
Chr.2L: 9616469 - 9623109 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032135
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169451.2 NP_001162922.1 4 - 5 1890 105
NM_135459.4 NP_609303.1 4 - 5 1659 105
NM_164864.2 NP_723476.1 4 - 5 930 105
GenBank mRNA AY058399.1 - 4 - 5 1637 105
BT133218.1 - 4 - 5 1806 105

View Full Product Details