Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nAChRalpha6
Entrez Gene ID
34304
Gene Name
nicotinic Acetylcholine Receptor alpha6
Gene Aliases
AChRalpha6, BcDNA:GH01410, CG4128, CT13662, Dalpha6, Dmel\CG4128, Dmel_CG4128, alpha6, das, nAChR, nAChR alpha 6, nAChR-alpha30D, nAChRAlpha-30D, nAChRalpha, nAChRalpha-30D, nAcRalpha-30D, rsn
Chromosome Location
Chr.2L: 9793317 - 9886250 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032151
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135472.6 NP_609316.4 9 - 10 2344 68
NM_164874.3 NP_723494.2 10 - 11 3268 68
NM_205952.3 NP_995674.1 11 - 12 3313 68
NM_205953.3 NP_995675.1 10 - 11 2225 68
GenBank mRNA AF321445.1 - 8 - 9 1275 68

View Full Product Details