Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bsk
Entrez Gene ID
44801
Gene Name
basket
Gene Aliases
BSK, Bsk, CG5680, D-JNK, D-junk, DBSK/JNK, DJNK, DJNK/bsk, DmJNK, Dmel\CG5680, Dmel_CG5680, JNK, JNK/SAPK, Jnk, Junk, P-JNK, SAPKa, dJNK, dJnk, dNJNK, jnk, jnk (bsk), p-JNK, pJNK
Chromosome Location
Chr.2L: 10247502 - 10250501 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000229
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169459.1 NP_001162930.1 4 - 5 1242 80
NM_001169461.3 NP_001162932.1 4 - 5 1100 80
NM_164900.3 NP_723541.1 5 - 6 980 80
GenBank mRNA AY070865.1 - 5 - 6 857 80
AY122221.1 - 5 - 6 959 80
U49180.1 - 4 - 5 1243 80

View Full Product Details