Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- salm
- Entrez Gene ID
-
34569
- Gene Name
- spalt major
- Gene Aliases
-
3602, B1164, CG6464, Dmel\CG6464, Dmel_CG6464, SAL, SALM, SAL[M], SPALT, Sal, SalM, Salm, Spalt, l(2)03602, sal, salII, salM, slam, spalt
- Chromosome Location
-
Chr.2L: 11434311 - 11445614 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261648
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_164966.3 |
NP_723670.2 |
3 - 4 |
4500 |
96 |
View Full Product Details