Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab3-GAP
Entrez Gene ID
34626
Gene Name
Rab3 GTPase activating protein
Gene Aliases
BcDNA:LD23336, CG7061, Dmel\CG7061, Dmel_CG7061, rab3-GAP
Chromosome Location
Chr.2L: 12058304 - 12063184 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027505
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135700.5 NP_609544.2 5 - 6 2555 65
NM_164989.2 NP_723718.2 5 - 6 2555 65
GenBank mRNA BT016089.1 - 5 - 6 2535 65

View Full Product Details