The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001273119.1 | NP_001260048.1 | 2 - 3 | 585 | 79 |
NM_135018.3 | NP_608862.1 | 2 - 3 | 585 | 79 | |
GenBank mRNA | AY118534.1 | - | 2 - 3 | 562 | 79 |