Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hoe1
Entrez Gene ID
249663
Gene Name
hoepel1
Gene Aliases
BcDNA:GH02636, CG11918, CG12787, CG2831, Dmel\CG12787, Dmel_CG12787, Hoe1, hoe, hoe-1
Chromosome Location
Chr.2L: 4908581 - 4932384 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041150
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273130.1 NP_001260059.1 10 - 11 3027 73
NM_001273131.1 NP_001260060.1 10 - 11 2955 73
NM_001298678.1 NP_001285607.1 9 - 10 2754 73
NM_135031.4 NP_608875.1 9 - 10 2739 73
NM_135032.2 NP_608876.1 9 - 10 2672 73
NM_164603.2 NP_723024.2 10 - 11 2883 73
NM_175966.2 NP_787980.1 10 - 11 2967 73
NM_175967.2 NP_787981.1 10 - 11 3005 73
NM_175968.2 NP_787982.1 10 - 11 2951 73
GenBank mRNA AF145605.1 - 9 - 10 2718 73
AY050571.1 - 10 - 11 2970 73
AY050572.1 - 9 - 10 2721 73
BT016131.1 - 10 - 11 2970 73
BT044574.1 - 9 - 10 2667 73
BT126019.1 - 7 - 8 2190 73

View Full Product Details