Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pgant5
- Entrez Gene ID
-
326151
- Gene Name
- Polypeptide N-Acetylgalactosaminyltransferase 5
- Gene Aliases
-
CG31651, CG9152, Dmel\CG31651, Dmel_CG31651, PGANT5, l(2)e02157, pgant5, pgnat5
- Chromosome Location
-
Chr.2L: 5072170 - 5093592 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031681
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135062.4 |
NP_608906.2 |
6 - 7 |
1948 |
75 |
View Full Product Details